Learn More
Grainger U-Bend HID Lamp Recycling Box
Catalog No. 502752368
Encompass_Preferred
The TerraCycle Regulated Waste U-Bend is a polyethylene U-bend that is designed for regulated waste applications. It features a 2" diameter and is made in the USA. This U-bend can be used with TerraCycle's regulated waste drums and pails to provide a secure seal and prevent leaks. It has a max temperature rating of 120°F and is compatible with corrosive chemicals. The dimensions are 3.5" x 2" x 2". This U-bend provides an economical and effective waste disposal solution for regulated waste streams.
Catalog No. 50-275-2368 Supplier Grainger Supplier No. 157F30
The TerraCycle Regulated Waste U-Bend is a polyethylene U-bend that is designed for regulated waste applications. It features a 2" diameter and is made in the USA. This U-bend can be used with TerraCycle's regulated waste drums and pails to provide a secure seal and prevent leaks. It has a max temperature rating of 120°F and is compatible with corrosive chemicals. The dimensions are 3.5" x 2" x 2". This U-bend provides an economical and effective waste disposal solution for regulated waste streams.