Learn More
Enterprise Technology Solutions U Brands Chalkboard, Magnetic, 48"Wx36"L, Chalkboard
SDP
Catalog No. 5000791676
Encompass_Preferred
Decor Magnetic Chalkboard provides a natural, contemporary surface. Rustic, 1-1/2" wide, decorative wood frame made of medium-density fiberboard adds durability and delivers a designer look. Simplified mounting hardware allows vertical or horizontal use. Actual measurements of the board surface are 47" wide x 35" high.
Catalog No. 50-007-91676 Supplier Enterprise Technology Solutions Supplier No. UBR4895U0001
Decor Magnetic Chalkboard provides a natural, contemporary surface. Rustic, 1-1/2" wide, decorative wood frame made of medium-density fiberboard adds durability and delivers a designer look. Simplified mounting hardware allows vertical or horizontal use. Actual measurements of the board surface are 47" wide x 35" high.