Learn More
Cayman Chemical U-ContoxInacetate 10mg
SDP
Catalog No. 502943923
Encompass_Preferred
A conotoxin and an inhibitor of Nav1.4 receptors; inhibits Nav1.4 in HEK293 cells (IC50 = 2.3 nM for the rat channel); inhibits rat Nav1.2 channels in HEK293 cells at 500 nM and is an antagonist of a7 nAChRs and a4ß2 and a3ß2 subunit-containing nAChRs in Xenopus oocytes expressing the human receptors (IC50s = 22, 2.9, and 0.45 µM, respectively); reduces global nerve action potential in isolated mouse sciatic nerves in a concentration-dependent manner; inhibits contraction in isolated mouse hemidiaphragm muscles (IC50 = 46 nM)
Catalog No. 50-294-3923 Supplier Cayman Chemical Supplier No. 4245510 MG
A conotoxin and an inhibitor of Nav1.4 receptors; inhibits Nav1.4 in HEK293 cells (IC50 = 2.3 nM for the rat channel); inhibits rat Nav1.2 channels in HEK293 cells at 500 nM and is an antagonist of a7 nAChRs and a4ß2 and a3ß2 subunit-containing nAChRs in Xenopus oocytes expressing the human receptors (IC50s = 22, 2.9, and 0.45 µM, respectively); reduces global nerve action potential in isolated mouse sciatic nerves in a concentration-dependent manner; inhibits contraction in isolated mouse hemidiaphragm muscles (IC50 = 46 nM)