Learn More
Ibidi USA standard µ-Dish 35mm, high. ESS 1.5KPA, 30 per box
SDP
Supplier:  Ibidi USA 81291
Encompass
A 35mm imaging dish with an elastically supported surface (ESS) used for growing cells under "in vivo-near" conditions. High resolution microscopy on soft cell culture surface based on the standard µ-Dish 35mm, high. Pcs./Box: 30 (individually packed)Elastic Surface ESS: ESS 1.5 kPaSurface Modification: ESS: uncoated, hydrophobic, sterilized
Catalog No. NC2029135