Learn More
Ibidi USA µ-Dish 35 mm, High, Glass Bottom - #1.5H Glass Bottom Coverslip
SDP
Catalog No. 50305807
Encompass
The ibidi product family is comprised of a variety of µ–Slides and µ–Dishes, which have all been designed for high–end microscopic analysis of fixed or living cells. The glass bottom versions of the µ–Slides and µ–Dishes are especially designed for TIRF, super resolution and single molecule applications. The µ-Dish 35 mm, high Glass Bottom allows you to perform high resolution microscopy in a 35 mm Petri dish with 12 mm walls. The standard height allows convenient liquid handling. The lid can be closed to reduce evaporation during long term experiments. Ø 21 mm observation area, #1.5H D 263 M Schott glass coverslip, uncoated, sterilized, 60 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-807 Supplier Ibidi USA Supplier No. 81158
The ibidi product family is comprised of a variety of µ–Slides and µ–Dishes, which have all been designed for high–end microscopic analysis of fixed or living cells. The glass bottom versions of the µ–Slides and µ–Dishes are especially designed for TIRF, super resolution and single molecule applications. The µ-Dish 35 mm, high Glass Bottom allows you to perform high resolution microscopy in a 35 mm Petri dish with 12 mm walls. The standard height allows convenient liquid handling. The lid can be closed to reduce evaporation during long term experiments. Ø 21 mm observation area, #1.5H D 263 M Schott glass coverslip, uncoated, sterilized, 60 pcs./box