Learn More
Ibidi USA µ-Dish 35 mm, High, Grid-500, ibiTreat - Tissue Culture Treated Polymer Coverslip
SDP
Catalog No. 50305809
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Dish 35 mm, high allows you to perform high resolution microscopy in a 35 mm Petri dish with 12 mm walls. The Grid–500 is a grid structure for relocating events. It provides 400 distinguishable observation squares of 500 µm edge length. The grid is clearly visible by phase contrast microscopy and based on the high quality ibidi Standard Bottom. The outer dimensions are identical to ibidi µ–Dishes. Ø 21 mm observation area, #1.5 polymer coverslip, tissue culture treated, sterilized, 60 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-809 Supplier Ibidi USA Supplier No. 81166
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Dish 35 mm, high allows you to perform high resolution microscopy in a 35 mm Petri dish with 12 mm walls. The Grid–500 is a grid structure for relocating events. It provides 400 distinguishable observation squares of 500 µm edge length. The grid is clearly visible by phase contrast microscopy and based on the high quality ibidi Standard Bottom. The outer dimensions are identical to ibidi µ–Dishes. Ø 21 mm observation area, #1.5 polymer coverslip, tissue culture treated, sterilized, 60 pcs./box