Learn More
Ibidi USA µ-Dish 35 mm, High, ibiTreat - Tissue Culture Treated Polymer Coverslip
SDP
Catalog No. 50305806
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Dish 35 mm, high allows you to perform high resolution microscopy in a 35 mm Petri dish with 12 mm walls. The standard height allows convenient liquid handling. The lid can be closed to reduce evaporation during long term experiments. Ø 21 mm observation area, #1.5 polymer coverslip, tissue culture treated, sterilized, 60 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-806 Supplier Ibidi USA Supplier No. 81156
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Dish 35 mm, high allows you to perform high resolution microscopy in a 35 mm Petri dish with 12 mm walls. The standard height allows convenient liquid handling. The lid can be closed to reduce evaporation during long term experiments. Ø 21 mm observation area, #1.5 polymer coverslip, tissue culture treated, sterilized, 60 pcs./box