Learn More
Ibidi USA U-DISH 35 mm LOW Glass Bottom #1.5h
SDP
Catalog No. NC2206010
Encompass
A 35mm imaging dish with a glass bottom and low walls for TIRF, single molecule and super-resolution microscopy. The dish contains a high-quality #1.5H Glass coverslip bottom with low thickness variability. Cell culture dish with low walls for a large access area during micromanipulation assays. Equipped with a tightly fitting lid that minimizes evaporation suitable for DIC when using a DIC LidSurface Modification: #1.5H (170 ¿m +/- 5 ¿m) D 263 M Schott glass, sterilizedPcs./Box: 60 (individually packed)
This item is not returnable. View return policy
Catalog No. NC2206010 Supplier Ibidi USA Supplier No. 80137
A 35mm imaging dish with a glass bottom and low walls for TIRF, single molecule and super-resolution microscopy. The dish contains a high-quality #1.5H Glass coverslip bottom with low thickness variability. Cell culture dish with low walls for a large access area during micromanipulation assays. Equipped with a tightly fitting lid that minimizes evaporation suitable for DIC when using a DIC LidSurface Modification: #1.5H (170 ¿m +/- 5 ¿m) D 263 M Schott glass, sterilizedPcs./Box: 60 (individually packed)