Learn More
Ibidi USA µ-Dish 35 mm, Low, Uncoated - Uncoated Polymer Coverslip
SDP
Catalog No. 50305743
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Dish 35mm, Low allows you to perform high resolution microscopy in a 35 mm Petri–dish with 7 mm walls. The low height makes high numerical apertures of Kohler illumination possible and provides large access for micromanipulation. The lid can be closed to hinder evaporation during long term experiments. Ø 35 mm, low wall (800 μl volume), #1.5 polymer coverslip, hydrophobic, sterilized, 60 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-743 Supplier Ibidi USA Supplier No. 80131
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Dish 35mm, Low allows you to perform high resolution microscopy in a 35 mm Petri–dish with 7 mm walls. The low height makes high numerical apertures of Kohler illumination possible and provides large access for micromanipulation. The lid can be closed to hinder evaporation during long term experiments. Ø 35 mm, low wall (800 μl volume), #1.5 polymer coverslip, hydrophobic, sterilized, 60 pcs./box