Learn More
Ibidi USA µ-Dish Microscopy Rack
SDP
Catalog No. 50305728
Encompass
µ-Dish Microscopy Rack is a aluminum rack made to hold 6 µ-Dishess for analysis of multiple samples. The multiwell format is compatible with all inverted microscopes. Magnetic Lids sold seperately. aluminum, black, 127.5 x 85.5 x 19.5 mm
This item is not returnable. View return policy
Catalog No. 50-305-728 Supplier Ibidi USA Supplier No. 80035
µ-Dish Microscopy Rack is a aluminum rack made to hold 6 µ-Dishess for analysis of multiple samples. The multiwell format is compatible with all inverted microscopes. Magnetic Lids sold seperately. aluminum, black, 127.5 x 85.5 x 19.5 mm