Learn More
Ibidi USA u-Pattern: RGD, 20 um squares, 110 um pitch, hexagonal 2/pk
SDP
Supplier:  Ibidi USA 83801S
Encompass
A micropatterned sufrace for single cell assays with fluorescence micrscopy readout. Two different singe-cell patterns with ideal spacing for various single cell assays. Allows for easy handling without preparation, unpack and get started! With excellent optical-quality imaging chamber for high-resolution micrscopy. Pcs./Box: 2 (Trial Pack), sterilized. µ-Pattern: RGD, 20 µm squares, 110 µm pitch, hexagonalSurface Modification: #1.5 polymer coverslip, micropatterned surface, surface passivation with BioinertProduct Variation: µ-Slide 8 Well high
Catalog No. NC2259127
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.