Learn More
Ibidi USA µ-Plate 96 Well Black, ibiTreat - Tissue Culture Treated Polymer Coverslip
SDP
Catalog No. 50305829
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, and µ-Plates which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Plate 96 Well Black allows you to perform high resolution microscopy in a standard multi-well format. The ibidi µ-Plate 96 Well Black is made out of a black polymer to eliminate well-to-well crosstalk during fluorescence microscopy. #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-829 Supplier Ibidi USA Supplier No. 89626
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, and µ-Plates which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Plate 96 Well Black allows you to perform high resolution microscopy in a standard multi-well format. The ibidi µ-Plate 96 Well Black is made out of a black polymer to eliminate well-to-well crosstalk during fluorescence microscopy. #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box