Learn More
Ibidi USA µ-Plate 96 Well Round Glass Bottom
SDP
Catalog No. NC2945536
Encompass
The ibidi product family is comprised of a variety of ¿-Slides and ¿-Dishes, and ¿-Plates which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The ¿-Plate 96 Well Black Round allows you to perform high resolution microscopy in a standard multi-well format. The ibidi ¿-Plate 96 Well Black is made out of a black polymer to eliminate well-to-well crosstalk during fluorescence microscopy. #1.5H (170 µm +/- 5 µm) D 263 M Schott glass, sterilized
Catalog No. NC2945536 Supplier Ibidi USA Supplier No. 89607
The ibidi product family is comprised of a variety of ¿-Slides and ¿-Dishes, and ¿-Plates which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The ¿-Plate 96 Well Black Round allows you to perform high resolution microscopy in a standard multi-well format. The ibidi ¿-Plate 96 Well Black is made out of a black polymer to eliminate well-to-well crosstalk during fluorescence microscopy. #1.5H (170 µm +/- 5 µm) D 263 M Schott glass, sterilized