Learn More
Meso Scale Discovery U-PLEX CAR-T Cell Combo 2 (human), kit, 7 analytes, electrochemiluminescence detection, low pg/mL detection limit, MULTI-ARRAY technology, human samples
SDP
Catalog No. NC3666205
Encompass
U-PLEX CAR-T Cell Combo 2 (human), kit, 7 analytes, electrochemiluminescence detection, low pg/mL detection limit, MULTI-ARRAY technology, human samples
Catalog No. NC3666205 Supplier Meso Scale Discovery Supplier No. K15600K2
Add to Cart
Add to Cart
Request a Quote
U-PLEX CAR-T Cell Combo 2 (human), kit, 7 analytes, electrochemiluminescence detection, low pg/mL detection limit, MULTI-ARRAY technology, human samples