Learn More
Meso Scale Discovery U-PLEX Chemokine Combo 1 hu
SDP
Catalog No. 5000081756
Encompass
The Meso Scale Discovery U-PLEX Chemokine Combo 1 Human Kit is a multiplex immunoassay that simultaneously measures the levels of 10 human chemokines CCL2 CCL3 CCL4 CCL5 CCL11 CXCL8 CXCL9 CXCL10 CXCL11 and CXCL12 This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is designed for use with serum plasma and cell culture supernatant samples The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and wash buffer This assay provides high sensitivity a broad dynamic range and requires a low sample volume of 50 L
Catalog No. 50-000-81756 Supplier Meso Scale Discovery Supplier No. K15047K1
The Meso Scale Discovery U-PLEX Chemokine Combo 1 Human Kit is a multiplex immunoassay that simultaneously measures the levels of 10 human chemokines CCL2 CCL3 CCL4 CCL5 CCL11 CXCL8 CXCL9 CXCL10 CXCL11 and CXCL12 This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is designed for use with serum plasma and cell culture supernatant samples The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and wash buffer This assay provides high sensitivity a broad dynamic range and requires a low sample volume of 50 L