Learn More
Meso Scale Discovery U-PLEX Chemokine Combo 1 ms
SDP
Catalog No. 5000082164
Encompass
The U-PLEX Chemokine Combo 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay that simultaneously measures mouse CCL2 CCL3 CCL4 CCL5 CXCL1 CXCL2 CXCL5 CXCL9 CXCL10 and CXCL13 in serum plasma and cell culture supernatants This 10-plex assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and requires as little as 25 L of sample The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators
Catalog No. 50-000-82164 Supplier Meso Scale Discovery Supplier No. K15321K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Chemokine Combo 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay that simultaneously measures mouse CCL2 CCL3 CCL4 CCL5 CXCL1 CXCL2 CXCL5 CXCL9 CXCL10 and CXCL13 in serum plasma and cell culture supernatants This 10-plex assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and requires as little as 25 L of sample The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators