Learn More
Meso Scale Discovery U-PLEX Chemokine Combo 2 hu
SDP
Catalog No. 5000079188
Encompass
The Meso Scale Discovery U-PLEX Chemokine Combo 2 Human Kit is a multiplex immunoassay that simultaneously measures the levels of 10 human chemokines in a single small sample volume The kit includes a 96-well plate pre-coated with capture antibodies for CCL2 CCL3 CCL4 CCL5 CCL11 CXCL1 CXCL8 CXCL9 CXCL10 and CXCL12 The kit also includes detection antibodies and calibrators to generate a standard curve The electrochemiluminescent assay takes approximately 5 hours to complete and only requires 50 L of sample per well The kit can be used to measure chemokines in serum plasma cell culture supernatants and other biological sample types
Catalog No. 50-000-79188 Supplier Meso Scale Discovery Supplier No. K15046K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Chemokine Combo 2 Human Kit is a multiplex immunoassay that simultaneously measures the levels of 10 human chemokines in a single small sample volume The kit includes a 96-well plate pre-coated with capture antibodies for CCL2 CCL3 CCL4 CCL5 CCL11 CXCL1 CXCL8 CXCL9 CXCL10 and CXCL12 The kit also includes detection antibodies and calibrators to generate a standard curve The electrochemiluminescent assay takes approximately 5 hours to complete and only requires 50 L of sample per well The kit can be used to measure chemokines in serum plasma cell culture supernatants and other biological sample types