Learn More
Meso Scale Discovery U-PLEX Chemokine Combo ms
SDP
Supplier:  Meso Scale Discovery K15099K1
Encompass
The U-PLEX Chemokine Combo Mouse Assay is a multiplex immunoassay panel for the simultaneous quantitative measurement of mouse chemokines CCL2 CCL3 CCL4 CCL5 CXCL1 CXCL2 CXCL5 CXCL9 CXCL10 and CXCL13 in cell culture supernatants serum plasma and other biological fluids This assay utilizes Meso Scale Discovery s electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The 10-plex panel allows efficient quantification of multiple chemokines from a single small sample volume
Catalog No. 50-000-79230