Learn More
Meso Scale Discovery U-PLEX Custom Biomarker Assay, Mouse, 96-well plate, Multiplex Immunoassay, Up to 10 analytes, Detection via ECL
SDP
Catalog No. NC3736093
Encompass
U-PLEX Custom Biomarker Assay, Mouse, 96-well plate, Multiplex Immunoassay, Up to 10 analytes, Detection via ECL
Catalog No. NC3736093 Supplier Meso Scale Discovery Supplier No. K15069M1
U-PLEX Custom Biomarker Assay, Mouse, 96-well plate, Multiplex Immunoassay, Up to 10 analytes, Detection via ECL