Learn More
Meso Scale Discovery U-PLEX Cytokine Combo 1 hu
SDP
Catalog No. 5000081821
Encompass
The Meso Scale Discovery U-PLEX Cytokine Combo 1 Human Kit (K15045K) is a multiplex immunoassay measuring 10 human cytokines and chemokines (IL-2 IL-4 IL-6 IL-10 IL-12p70 IL-13 IFN- TNF- GM-CSF CCL2) from various samples Using MSD s MULTI-ARRAY technology it runs on the MESO QuickPlex SQ 120 The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and reagents Results are available in 3 5 hours offering high sensitivity broad range and low sample volume
Catalog No. 50-000-81821 Supplier Meso Scale Discovery Supplier No. K15045K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Cytokine Combo 1 Human Kit (K15045K) is a multiplex immunoassay measuring 10 human cytokines and chemokines (IL-2 IL-4 IL-6 IL-10 IL-12p70 IL-13 IFN- TNF- GM-CSF CCL2) from various samples Using MSD s MULTI-ARRAY technology it runs on the MESO QuickPlex SQ 120 The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and reagents Results are available in 3 5 hours offering high sensitivity broad range and low sample volume