Learn More
Meso Scale Discovery U-PLEX Cytokine Combo 1 NHP
SDP
Catalog No. 5000078278
Encompass
The U-PLEX Cytokine Combo 1 NHP Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring 15 non-human primate cytokines chemokines and growth factors (e g IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12/23p40 IL-13 IL-17A/F IFN- TNF- GM-CSF CCL2/MCP-1 CXCL10/IP-10 VEGF-A) in serum plasma and culture supernatants It uses electrochemiluminescence detection on the MESO QuickPlex SQ 120 The kit includes a 96-well plate antibodies buffers diluent calibrators and controls enabling high-throughput sensitive and reproducible quantification from small samples
Catalog No. 50-000-78278 Supplier Meso Scale Discovery Supplier No. K15057K1
The U-PLEX Cytokine Combo 1 NHP Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring 15 non-human primate cytokines chemokines and growth factors (e g IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12/23p40 IL-13 IL-17A/F IFN- TNF- GM-CSF CCL2/MCP-1 CXCL10/IP-10 VEGF-A) in serum plasma and culture supernatants It uses electrochemiluminescence detection on the MESO QuickPlex SQ 120 The kit includes a 96-well plate antibodies buffers diluent calibrators and controls enabling high-throughput sensitive and reproducible quantification from small samples