Learn More
Meso Scale Discovery U-PLEX Cytokine Combo 1 NHP
SDP
Catalog No. 5000081675
Encompass
The Meso Scale Discovery U-PLEX Cytokine Combo 1 NHP Kit (K15057K) is a multiplex immunoassay kit for the simultaneous detection of 15 non-human primate cytokines and chemokines in serum plasma and cell culture supernatants The kit uses electrochemiluminescence technology on patterned arrays to provide sensitive and accurate results from small sample volumes The 15 analytes measured are Eotaxin G-CSF GM-CSF IFN- IL-1 IL-1ra IL-2 IL-4 IL-5 IL-6 IL-8 IL-10 IL-12/23p40 MCP-1 and TNF- The kit includes a 96-well plate calibrators controls detection antibodies read buffer wash buffer and other assay components for 96 tests
Catalog No. 50-000-81675 Supplier Meso Scale Discovery Supplier No. K15057K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Cytokine Combo 1 NHP Kit (K15057K) is a multiplex immunoassay kit for the simultaneous detection of 15 non-human primate cytokines and chemokines in serum plasma and cell culture supernatants The kit uses electrochemiluminescence technology on patterned arrays to provide sensitive and accurate results from small sample volumes The 15 analytes measured are Eotaxin G-CSF GM-CSF IFN- IL-1 IL-1ra IL-2 IL-4 IL-5 IL-6 IL-8 IL-10 IL-12/23p40 MCP-1 and TNF- The kit includes a 96-well plate calibrators controls detection antibodies read buffer wash buffer and other assay components for 96 tests