Learn More
Meso Scale Discovery U-PLEX DEVELOPMENT PK,4-ASY 25
SDP
Catalog No. NC1854887
Encompass
U-PLEX Development Pack,4-Assay - 25 plates.The pack contains a 10-spot U-PLEX plate with 4 activated spots, 4 unique linkers, and all the necessary diluents and buffers to complete the assays.
Catalog No. NC1854887 Supplier Meso Scale Discovery Supplier No. K15229N4
Add to Cart
Add to Cart
Request a Quote
U-PLEX Development Pack,4-Assay - 25 plates.The pack contains a 10-spot U-PLEX plate with 4 activated spots, 4 unique linkers, and all the necessary diluents and buffers to complete the assays.