Learn More
Meso Scale Discovery U-PLEX Human A2M ASY 25 PL
SDP
Catalog No. 5000078790
Encompass
The Meso Scale Discovery U-PLEX Human A2M Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Alpha-2-Macroglobulin (A2M) in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and accurate detection of A2M levels down to sub-pg/mL concentrations The assay requires 50 L sample volume and can be read on MSD instrument platforms This product provides a simple rapid way to quantify A2M levels which can aid research into inflammatory conditions immune function and proteolytic regulation
Catalog No. 50-000-78790 Supplier Meso Scale Discovery Supplier No. K151Q9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human A2M Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Alpha-2-Macroglobulin (A2M) in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and accurate detection of A2M levels down to sub-pg/mL concentrations The assay requires 50 L sample volume and can be read on MSD instrument platforms This product provides a simple rapid way to quantify A2M levels which can aid research into inflammatory conditions immune function and proteolytic regulation