Learn More
Meso Scale Discovery U-PLEX Human A2M ASY
SDP
Catalog No. 5000078577
Encompass
The Meso Scale Discovery U-PLEX Human A2M Assay is a multiplex immunoassay kit for measuring Alpha-2-Macroglobulin (A2M) in human serum plasma and cell culture supernatant This electrochemiluminescent assay uses MSD s SECTOR Imager offering high sensitivity a broad dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls It enables rapid reproducible quantification of A2M crucial for regulating protease activity
Catalog No. 50-000-78577 Supplier Meso Scale Discovery Supplier No. K151Q9K2
The Meso Scale Discovery U-PLEX Human A2M Assay is a multiplex immunoassay kit for measuring Alpha-2-Macroglobulin (A2M) in human serum plasma and cell culture supernatant This electrochemiluminescent assay uses MSD s SECTOR Imager offering high sensitivity a broad dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls It enables rapid reproducible quantification of A2M crucial for regulating protease activity