Learn More
Meso Scale Discovery U-PLEX Human A2M ASY
SDP
Catalog No. 5000079838
Encompass
The Meso Scale Discovery U-PLEX Human A2M Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Alpha-2-Macroglobulin (A2M) in human serum plasma and cell culture supernatant samples This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay allows rapid reproducible quantification of A2M which plays a key role in regulating protease activity
Catalog No. 50-000-79838 Supplier Meso Scale Discovery Supplier No. K151Q9K1
The Meso Scale Discovery U-PLEX Human A2M Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Alpha-2-Macroglobulin (A2M) in human serum plasma and cell culture supernatant samples This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay allows rapid reproducible quantification of A2M which plays a key role in regulating protease activity