Learn More
Meso Scale Discovery U-PLEX Human A2M ASY384-well
SDP
Catalog No. 5000081062
Encompass
The Meso Scale Discovery U-PLEX Human A2M 384-Well Assay is a multiplex immunoassay that measures A2M in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This 384-well plate assay allows for the simultaneous measurement of A2M along with other biomarkers It only requires 10 L of sample per well and the entire workflow can be completed in around 3 hours This assay kit includes a 384-well MULTI-ARRAY Plate Detection Antibody Read Buffer Calibrator and Diluent
Catalog No. 50-000-81062 Supplier Meso Scale Discovery Supplier No. K251Q9K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human A2M 384-Well Assay is a multiplex immunoassay that measures A2M in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This 384-well plate assay allows for the simultaneous measurement of A2M along with other biomarkers It only requires 10 L of sample per well and the entire workflow can be completed in around 3 hours This assay kit includes a 384-well MULTI-ARRAY Plate Detection Antibody Read Buffer Calibrator and Diluent