Learn More
Meso Scale Discovery U-PLEX Human A2M ASYQP
SDP
Catalog No. 5000079840
Encompass
The Meso Scale Discovery U-PLEX Human A2M Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Alpha-2-Macroglobulin (A2M) in human serum plasma and cell culture supernatant samples This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve Results are obtained in as little as 3 5 hours This assay is for research use only
Catalog No. 50-000-79840 Supplier Meso Scale Discovery Supplier No. K151Q9K22
The Meso Scale Discovery U-PLEX Human A2M Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Alpha-2-Macroglobulin (A2M) in human serum plasma and cell culture supernatant samples This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve Results are obtained in as little as 3 5 hours This assay is for research use only