Learn More
Meso Scale Discovery U-PLEX Human Adiponectin Assay, K151R9K
SDP
Catalog No. NC2227620
Encompass
The Meso Scale Discovery U-PLEX Human Adiponectin Assay Kit is designed for the quantitative measurement of adiponectin in human serum, plasma, and cell culture supernatants using an electrochemiluminescent detection method. The assay uses a plate coated with capture antibodies and U-PLEX linker conjugated detection antibodies specific for adiponectin. It has a detection range of 50-10,000 pg/mL and includes calibrators and controls. The kit includes a 96-well plate, detection antibody, read buffer, wash buffer, sample diluent, calibrators, controls, and plate seals. It allows high sensitivity, low sample volume, and short assay time.
This item is not returnable. View return policy
Catalog No. NC2227620 Supplier Meso Scale Discovery Supplier No. K151R9K2
The Meso Scale Discovery U-PLEX Human Adiponectin Assay Kit is designed for the quantitative measurement of adiponectin in human serum, plasma, and cell culture supernatants using an electrochemiluminescent detection method. The assay uses a plate coated with capture antibodies and U-PLEX linker conjugated detection antibodies specific for adiponectin. It has a detection range of 50-10,000 pg/mL and includes calibrators and controls. The kit includes a 96-well plate, detection antibody, read buffer, wash buffer, sample diluent, calibrators, controls, and plate seals. It allows high sensitivity, low sample volume, and short assay time.