Learn More
Meso Scale Discovery U-PLEX Human Adiponectin ASY
SDP
Catalog No. 5000082196
Encompass
The Meso Scale Discovery U-PLEX Human Adiponectin Assay Kit quantifies adiponectin in human serum plasma and cell culture supernatants using electrochemiluminescent detection It features MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and minimal interference The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents The assay takes about 3 5 hours facilitating adiponectin measurement in studies on obesity diabetes and cardiovascular disease
Catalog No. 50-000-82196 Supplier Meso Scale Discovery Supplier No. K151R9K1
The Meso Scale Discovery U-PLEX Human Adiponectin Assay Kit quantifies adiponectin in human serum plasma and cell culture supernatants using electrochemiluminescent detection It features MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and minimal interference The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents The assay takes about 3 5 hours facilitating adiponectin measurement in studies on obesity diabetes and cardiovascular disease