Learn More
Meso Scale Discovery U-PLEX Human ApoA1 ASY 25 PL
SDP
Catalog No. 5000082307
Encompass
The Meso Scale Discovery U-PLEX Human ApoA1 Assay Kit is a multiplex immunoassay that measures Apolipoprotein A1 (ApoA1) levels in human serum plasma and cell culture supernatant samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator and controls The assay has a typical sensitivity of 0 3 ng/mL and an upper limit of quantification of 2500 ng/mL This product can aid in biomarker research related to cardiovascular disease lipid metabolism and inflammation
Catalog No. 50-000-82307 Supplier Meso Scale Discovery Supplier No. K151S9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ApoA1 Assay Kit is a multiplex immunoassay that measures Apolipoprotein A1 (ApoA1) levels in human serum plasma and cell culture supernatant samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator and controls The assay has a typical sensitivity of 0 3 ng/mL and an upper limit of quantification of 2500 ng/mL This product can aid in biomarker research related to cardiovascular disease lipid metabolism and inflammation