Learn More
Meso Scale Discovery U-PLEX Human ApoA1 ASY
SDP
Catalog No. 5000078563
Encompass
The Meso Scale Discovery U-PLEX Human ApoA1 Assay Kit is a multiplex immunoassay that measures Apolipoprotein A1 (ApoA1) levels in human serum plasma and cell culture supernatant samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator This quantitative immunoassay allows users to accurately determine ApoA1 concentrations within 90 minutes using as little as 25 L of sample
Catalog No. 50-000-78563 Supplier Meso Scale Discovery Supplier No. K151S9K1
The Meso Scale Discovery U-PLEX Human ApoA1 Assay Kit is a multiplex immunoassay that measures Apolipoprotein A1 (ApoA1) levels in human serum plasma and cell culture supernatant samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator This quantitative immunoassay allows users to accurately determine ApoA1 concentrations within 90 minutes using as little as 25 L of sample