Learn More
Meso Scale Discovery U-PLEX Human ApoA1 ASY
SDP
Catalog No. 5000081298
Encompass
The Meso Scale Discovery U-PLEX Human ApoA1 Assay Kit is a multiplex immunoassay for measuring Apolipoprotein A1 (ApoA1) in human serum plasma and cell culture supernatant It uses electrochemiluminescence (ECL) detection for high sensitivity and rapid results Requiring 50 L of sample it is compatible with Meso Scale Discovery SECTOR Imager instruments The kit includes a 96-well plate with capture and detection antibodies Read Buffer Diluent 2 and calibrator enabling quantitative measurement of ApoA1 a key component of HDL involved in cholesterol transport
Catalog No. 50-000-81298 Supplier Meso Scale Discovery Supplier No. K151S9K2
The Meso Scale Discovery U-PLEX Human ApoA1 Assay Kit is a multiplex immunoassay for measuring Apolipoprotein A1 (ApoA1) in human serum plasma and cell culture supernatant It uses electrochemiluminescence (ECL) detection for high sensitivity and rapid results Requiring 50 L of sample it is compatible with Meso Scale Discovery SECTOR Imager instruments The kit includes a 96-well plate with capture and detection antibodies Read Buffer Diluent 2 and calibrator enabling quantitative measurement of ApoA1 a key component of HDL involved in cholesterol transport