Learn More
Meso Scale Discovery U-PLEX Human ApoA1 ASYQP
SDP
Catalog No. 5000080545
Encompass
The Meso Scale Discovery U-PLEX Human ApoA1 Assay Kit quantifies Apolipoprotein A1 (ApoA1) in human serum plasma and cell culture supernatants using electrochemiluminescent detection A capture antibody specific for ApoA1 coats the plate Standards and samples are added binding ApoA1 to the antibody After washing a detection antibody with an electrochemiluminescent label is added Light emission upon stimulation indicates analyte levels The kit includes a 96-well plate detection antibody standards buffers substrate and seals
Catalog No. 50-000-80545 Supplier Meso Scale Discovery Supplier No. K151S9K21
The Meso Scale Discovery U-PLEX Human ApoA1 Assay Kit quantifies Apolipoprotein A1 (ApoA1) in human serum plasma and cell culture supernatants using electrochemiluminescent detection A capture antibody specific for ApoA1 coats the plate Standards and samples are added binding ApoA1 to the antibody After washing a detection antibody with an electrochemiluminescent label is added Light emission upon stimulation indicates analyte levels The kit includes a 96-well plate detection antibody standards buffers substrate and seals