Learn More
Meso Scale Discovery U-PLEX Human ApoA1 ASYQP
SDP
Catalog No. 5000080548
Encompass
The Meso Scale Discovery U-PLEX Human ApoA1 Assay Kit measures Apolipoprotein A1 (ApoA1) in human serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results It includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay offers a quick way to measure ApoA1 levels in various human samples
Catalog No. 50-000-80548 Supplier Meso Scale Discovery Supplier No. K151S9K22
The Meso Scale Discovery U-PLEX Human ApoA1 Assay Kit measures Apolipoprotein A1 (ApoA1) in human serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results It includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay offers a quick way to measure ApoA1 levels in various human samples