Learn More
Meso Scale Discovery U-PLEX Human ApoC3 Assay, Immunoassay, 4 plate size, Dynamic Range 1.06 - 10,000 pg/mL, Lower Limit of Detection 1.06 pg/mL, 96-well SECTOR plates, For research use only
SDP
Catalog No. NC3257489
Encompass
The MESO SCALE DISCOVERY U-PLEX Human ApoC3 Assay is an advanced immunoassay for the precise quantitative detection of human Apolipoprotein C-III (ApoC3). This assay is designed for use with various sample types and is suitable for metabolic research applications, providing high sensitivity and specificity. Key features include compatibility with multiple MSD instrument systems and minimal non-specific binding.
  • Precise measurement of ApoC3
  • High sensitivity with a lower limit of detection of 1.06 pg/mL
  • Wide dynamic range from 1.06 to 10,000 pg/mL
  • Multiple sample type compatibility including serum, plasma, and cell culture media
  • Supports multiplexing options
  • Less than 0.5% non-specific binding
  • Uses mouse monoclonal capture and detection antibodies
  • For research use only
  • Stored as multi-component kit
  • Fits with Biomarker Group 3
Catalog No. NC3257489 Supplier Meso Scale Discovery Supplier No. K151T9K2
The MESO SCALE DISCOVERY U-PLEX Human ApoC3 Assay is an advanced immunoassay for the precise quantitative detection of human Apolipoprotein C-III (ApoC3). This assay is designed for use with various sample types and is suitable for metabolic research applications, providing high sensitivity and specificity. Key features include compatibility with multiple MSD instrument systems and minimal non-specific binding.
  • Precise measurement of ApoC3
  • High sensitivity with a lower limit of detection of 1.06 pg/mL
  • Wide dynamic range from 1.06 to 10,000 pg/mL
  • Multiple sample type compatibility including serum, plasma, and cell culture media
  • Supports multiplexing options
  • Less than 0.5% non-specific binding
  • Uses mouse monoclonal capture and detection antibodies
  • For research use only
  • Stored as multi-component kit
  • Fits with Biomarker Group 3