Learn More
Meso Scale Discovery U-PLEX Human ApoC3 ASY 25 PL
SDP
Catalog No. 5000079832
Encompass
The Meso Scale Discovery U-PLEX Human ApoC3 Assay Kit is a multiplex immunoassay that measures Apolipoprotein C3 (ApoC3) levels in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for the quantitative measurement of human ApoC3 This assay allows for the rapid reproducible quantification of ApoC3 levels which can aid in cardiovascular disease and metabolic disorder research
Catalog No. 50-000-79832 Supplier Meso Scale Discovery Supplier No. K151T9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ApoC3 Assay Kit is a multiplex immunoassay that measures Apolipoprotein C3 (ApoC3) levels in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for the quantitative measurement of human ApoC3 This assay allows for the rapid reproducible quantification of ApoC3 levels which can aid in cardiovascular disease and metabolic disorder research