Learn More
Meso Scale Discovery U-PLEX Human ApoC3 ASY
SDP
Catalog No. 5000079167
Encompass
The Meso Scale Discovery U-PLEX Human ApoC3 Assay is an electrochemiluminescent immunoassay for measuring Apolipoprotein C3 (ApoC3) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of ApoC3 a key regulator of triglyceride metabolism The assay detects from 0 3 ng/mL to 10 000 ng/mL requires 50 L of sample and is read on MSD s QuickPlex SQ 120 It helps researchers quantify ApoC3 for studying dyslipidemias cardiovascular disease and metabolic disorders
Catalog No. 50-000-79167 Supplier Meso Scale Discovery Supplier No. K151T9K1
The Meso Scale Discovery U-PLEX Human ApoC3 Assay is an electrochemiluminescent immunoassay for measuring Apolipoprotein C3 (ApoC3) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of ApoC3 a key regulator of triglyceride metabolism The assay detects from 0 3 ng/mL to 10 000 ng/mL requires 50 L of sample and is read on MSD s QuickPlex SQ 120 It helps researchers quantify ApoC3 for studying dyslipidemias cardiovascular disease and metabolic disorders