Learn More
Meso Scale Discovery U-PLEX Human ApoC3 ASY384-well
SDP
Catalog No. 5000079863
Encompass
The Meso Scale Discovery U-PLEX Human ApoC3 384-well Assay is an electrochemiluminescent immunoassay designed for the quantitative measurement of Apolipoprotein C3 (ApoC3) in human serum plasma and cell culture supernatants This 384-well plate assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of ApoC3 an important regulator of triglyceride metabolism With a sample volume of 25 L and an assay time of 2 5 hours this assay offers high-throughput capabilities ideal for biomarker research and drug discovery applications
Catalog No. 50-000-79863 Supplier Meso Scale Discovery Supplier No. K251T9K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ApoC3 384-well Assay is an electrochemiluminescent immunoassay designed for the quantitative measurement of Apolipoprotein C3 (ApoC3) in human serum plasma and cell culture supernatants This 384-well plate assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of ApoC3 an important regulator of triglyceride metabolism With a sample volume of 25 L and an assay time of 2 5 hours this assay offers high-throughput capabilities ideal for biomarker research and drug discovery applications