Learn More
Meso Scale Discovery U-PLEX Human ApoC3 ASYQP
SDP
Catalog No. 5000081358
Encompass
The Meso Scale Discovery U-PLEX Human ApoC3 Assay Kit measures Apolipoprotein C3 (ApoC3) in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of ApoC3 down to the pg/mL range It requires a small sample volume and offers high-throughput capabilities for research on lipid metabolism and related disorders The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls buffers and seals for one plate
Catalog No. 50-000-81358 Supplier Meso Scale Discovery Supplier No. K151T9K21
The Meso Scale Discovery U-PLEX Human ApoC3 Assay Kit measures Apolipoprotein C3 (ApoC3) in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of ApoC3 down to the pg/mL range It requires a small sample volume and offers high-throughput capabilities for research on lipid metabolism and related disorders The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls buffers and seals for one plate