Learn More
Meso Scale Discovery U-PLEX Human ApoC3 ASYQP
SDP
Catalog No. 5000082303
Encompass
The Meso Scale Discovery U-PLEX Human ApoC3 Assay Kit is a multiplex immunoassay that measures Apolipoprotein C3 (ApoC3) levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls Results are obtained in as little as 3 5 hours using an MSD instrument This assay allows for the quantitative measurement of ApoC3 an important regulator of triglyceride metabolism
Catalog No. 50-000-82303 Supplier Meso Scale Discovery Supplier No. K151T9K22
The Meso Scale Discovery U-PLEX Human ApoC3 Assay Kit is a multiplex immunoassay that measures Apolipoprotein C3 (ApoC3) levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls Results are obtained in as little as 3 5 hours using an MSD instrument This assay allows for the quantitative measurement of ApoC3 an important regulator of triglyceride metabolism