Learn More
Meso Scale Discovery U-PLEX HUMAN B-NGF ASSAY, 5 plates, 96-well format, Sensitivity 0.052 pg/mL, Dynamic Range 0.052 - 498 pg/mL, Serum, EDTA Plasma, P800 Plasma
SDP
Catalog No. NC3368347
Encompass
U-PLEX HUMAN B-NGF ASSAY, 5 plates, 96-well format, Sensitivity 0.052 pg/mL, Dynamic Range 0.052 - 498 pg/mL, Serum, EDTA Plasma, P800 Plasma
Catalog No. NC3368347 Supplier Meso Scale Discovery Supplier No. K1516DK2
U-PLEX HUMAN B-NGF ASSAY, 5 plates, 96-well format, Sensitivity 0.052 pg/mL, Dynamic Range 0.052 - 498 pg/mL, Serum, EDTA Plasma, P800 Plasma