Learn More
Meso Scale Discovery U-PLEX Human B-NGF ASY384-well
SDP
Catalog No. 5000078360
Encompass
The Meso Scale Discovery U-PLEX Human Beta-NGF 384-Well Assay is a multiplex immunoassay panel that measures the neurotrophin Beta-NGF in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology in a 384-well plate format providing high sensitivity and a broad dynamic range This assay allows for the quantitative measurement of Beta-NGF with high precision and reproducibility It requires a low sample volume of 25 L and the total assay time is around 3 5 hours This product can aid in understanding Beta-NGF biology and pathology in areas such as neuroscience immunology oncology and pain
Catalog No. 50-000-78360 Supplier Meso Scale Discovery Supplier No. K2516DK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Beta-NGF 384-Well Assay is a multiplex immunoassay panel that measures the neurotrophin Beta-NGF in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology in a 384-well plate format providing high sensitivity and a broad dynamic range This assay allows for the quantitative measurement of Beta-NGF with high precision and reproducibility It requires a low sample volume of 25 L and the total assay time is around 3 5 hours This product can aid in understanding Beta-NGF biology and pathology in areas such as neuroscience immunology oncology and pain