Learn More
Meso Scale Discovery U-PLEX Human BCMA/TNFRSF17 ASY
SDP
Catalog No. 5000082057
Encompass
The Meso Scale Discovery U-PLEX Human BCMA (TNFRSF17) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of BCMA in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The assay has a lower limit of detection of 19 pg/mL and includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for BCMA quantification This multiplex assay enables the simultaneous measurement of BCMA along with other biomarkers It is suitable for research use only
Catalog No. 50-000-82057 Supplier Meso Scale Discovery Supplier No. K151D8K2
The Meso Scale Discovery U-PLEX Human BCMA (TNFRSF17) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of BCMA in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The assay has a lower limit of detection of 19 pg/mL and includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for BCMA quantification This multiplex assay enables the simultaneous measurement of BCMA along with other biomarkers It is suitable for research use only