Learn More
Meso Scale Discovery U-PLEX Human BCMA/TNFRSF17 ASY
SDP
Catalog No. 5000082060
Encompass
The Meso Scale Discovery U-PLEX Human BCMA (TNFRSF17) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of BCMA in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results for BCMA levels between 0 156 pg/mL to 10 000 pg/mL This assay can be used to monitor BCMA levels in research related to multiple myeloma autoimmune diseases and immune system function The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X read buffer sample diluent and plate seals
Catalog No. 50-000-82060 Supplier Meso Scale Discovery Supplier No. K151D8K1
The Meso Scale Discovery U-PLEX Human BCMA (TNFRSF17) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of BCMA in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results for BCMA levels between 0 156 pg/mL to 10 000 pg/mL This assay can be used to monitor BCMA levels in research related to multiple myeloma autoimmune diseases and immune system function The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X read buffer sample diluent and plate seals