Learn More
Meso Scale Discovery U-PLEX Human CA1 ASY 25 PL
SDP
Catalog No. 5000081174
Encompass
The Meso Scale Discovery U-PLEX Human CA1 Assay is a multiplex immunoassay kit for measuring CA1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results Requiring 50 L or less sample volume it is compatible with Meso Scale Discovery platforms The kit includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation and assay validation offering a simple sensitive and reproducible method to measure CA1 levels
Catalog No. 50-000-81174 Supplier Meso Scale Discovery Supplier No. K151J9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CA1 Assay is a multiplex immunoassay kit for measuring CA1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results Requiring 50 L or less sample volume it is compatible with Meso Scale Discovery platforms The kit includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation and assay validation offering a simple sensitive and reproducible method to measure CA1 levels