Learn More
Meso Scale Discovery U-PLEX Human CA1 ASY
SDP
Catalog No. 5000079277
Encompass
The Meso Scale Discovery U-PLEX Human CA1 Assay is a multiplex immunoassay kit for measuring CA1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and diluent It quantifies CA1 levels with minimal hands-on time using an MSD instrument completing tests in 3 5 hours for efficient CA1 biomarker analysis
Catalog No. 50-000-79277 Supplier Meso Scale Discovery Supplier No. K151J9K2
The Meso Scale Discovery U-PLEX Human CA1 Assay is a multiplex immunoassay kit for measuring CA1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and diluent It quantifies CA1 levels with minimal hands-on time using an MSD instrument completing tests in 3 5 hours for efficient CA1 biomarker analysis