Learn More
Meso Scale Discovery U-PLEX Human CA1 ASY
SDP
Catalog No. 5000081152
Encompass
The Meso Scale Discovery U-PLEX Human CA1 Assay is a multiplex immunoassay kit for measuring CA1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The assay requires 50 L or less sample volume and is compatible with Meso Scale Discovery platforms It includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay efficiently quantifies CA1 important in cancer and inflammatory diseases
Catalog No. 50-000-81152 Supplier Meso Scale Discovery Supplier No. K151J9K1
The Meso Scale Discovery U-PLEX Human CA1 Assay is a multiplex immunoassay kit for measuring CA1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The assay requires 50 L or less sample volume and is compatible with Meso Scale Discovery platforms It includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay efficiently quantifies CA1 important in cancer and inflammatory diseases