Learn More
Meso Scale Discovery U-PLEX Human CA1 ASY384-well
SDP
Catalog No. 5000080655
Encompass
The U-PLEX Human CA1 384-Well Assay from Meso Scale Diagnostics is a 384-well electrochemiluminescent immunoassay for measuring CA1 in human serum plasma (EDTA heparin) and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for a rapid ultra-sensitive cost-effective method with a detection limit of 0 03 ng/mL and quantification up to 1500 ng/mL Requiring 10 L per well it performs up to 40 tests on two plates offering a high-throughput reproducible immunoassay for CA1 research
Catalog No. 50-000-80655 Supplier Meso Scale Discovery Supplier No. K251J9K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human CA1 384-Well Assay from Meso Scale Diagnostics is a 384-well electrochemiluminescent immunoassay for measuring CA1 in human serum plasma (EDTA heparin) and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for a rapid ultra-sensitive cost-effective method with a detection limit of 0 03 ng/mL and quantification up to 1500 ng/mL Requiring 10 L per well it performs up to 40 tests on two plates offering a high-throughput reproducible immunoassay for CA1 research