Learn More
Meso Scale Discovery U-PLEX Human CA1 ASYQP 25 PL
SDP
Catalog No. 5000079928
Encompass
The Meso Scale Discovery U-PLEX Human CA1 Assay is a multiplex immunoassay kit for quantifying CA1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results Requiring 50 L or less sample volume it is compatible with Meso Scale Discovery platforms The kit includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation and assay validation It enables sensitive reproducible CA1 measurement aiding research on carbonic anhydrases and related diseases
Catalog No. 50-000-79928 Supplier Meso Scale Discovery Supplier No. K151J9K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CA1 Assay is a multiplex immunoassay kit for quantifying CA1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results Requiring 50 L or less sample volume it is compatible with Meso Scale Discovery platforms The kit includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation and assay validation It enables sensitive reproducible CA1 measurement aiding research on carbonic anhydrases and related diseases